missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Aqualine™ Titrant 5, for Karl Fischer Titration By Volumetry, Fisher Chemical

Catalog No. K22001
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K22001 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K22001 Supplier Fisher Chemical Supplier No. K220015
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For two-component system. Water equivalent: 5.0mg H2O/mL

Specifications

Chemical Name or Material Aqualine™ Titrant 5
Name Note Karl Fischer Reagent
Boiling Point 64°C
Color Colorless
Physical Form Liquid
Quantity 1 L
Molecular Formula CH4O
Packaging Amber glass bottle
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Show More Show Less