missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Aqualine™ Solvent CM, for Karl Fischer Titration By Volumetry, Fisher Chemical™

Catalog No. AL2110212
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AL2110212 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AL2110212 Supplier Thermo Fisher Scientific Supplier No. AL2110212
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Solvent for samples with high hydrocarbon content
  • Used in conjunction with Aqualine™ titrant for two component volumetric Karl Fischer analysis

Specifications

Chemical Name or Material Aqualine™ Solvent CM
Name Note Karl Fischer Reagent
CAS 288-32-4, 67-66-3, 7446-09-5
Physical Form Liquid
Quantity 2.5 L
Packaging Amber glass bottle
MDL Number 81734