missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Aqualine™ Sodium Tartrate-2H2O, for Karl Fischer Titration, Fisher Chemical

Catalog No. AL2770100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AL2770100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AL2770100 Supplier Thermo Fisher Scientific Supplier No. AL2770100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Solid standard

  • Primary solid water standard for Karl Fischer volumetric analysis
  • Contains 15.66% ± 0.05% water

Chemical Identifiers

Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
PubChem CID 131855972
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

Color White
pH 7 to 9
Physical Form Powder/Solid
CAS 6106-24-7
Quantity 100 g
Identification Pass Test
Packaging Plastic powder jar
Formula Weight 230.08g/mol
Chemical Name or Material Aqualine™ Sodium Tartrate Dihydrate Water Standard, Karl Fischer Reagent