missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Aqualine™ Electrolyte A, for Karl Fischer Titration By Coulometry, Fisher Chemical

Catalog No. K2500500
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K2500500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K2500500 Supplier Fisher Chemical Supplier No. K2500 08

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Anolyte solution for general use in conventional cells with a diaphragm

  • Anolyte solution for use in conventional cells with a diaphragm
  • Source of electrolyte required for anodic oxidation for coulometric analysis
  • Contains methanol and chloroform as solvents

Specifications

Chemical Name or Material Aqualine™ Electrolyte A
Name Note Karl Fischer Reagent
Color Colorless
CAS 108-75-8, 7446-09-5, 67-66-3, 111-42-2
Physical Form Liquid
Quantity 500 mL
Packaging Amber glass bottle
MDL Number 81734
UN Number 1992