Learn More
Aqualine™ Complete 2, Fisher Chemical™
Single-component reagent; Pyridine free
$233.90 - $565.48
Specifications
Physical Form Liquid
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Packaging Color Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Color Price Quantity  
K1950-1
View Documents
Fisher Chemical
K195015
1 L Amber Glass Colorless
Each for $233.90
Only null left
K1950-212
Accessories View Documents
Fisher Chemical
K195017
2.5 L Amber glass bottle Colorless
Each for $565.48
Only null left
Description
Description
  • Single-component volumetric reagent for high water content analysis
  • Water equivalent: 2.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free
Specifications
Specifications
Liquid