missing translation for 'onlineSavingsMsg'
Learn More

Aqualine™ Complete 2, Fisher Chemical™

Catalog No. K1950212
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass
Amber glass bottle
Color:
Colorless
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Color
K1950212 2.5 L Amber glass bottle Colorless
K19501 1 L Amber Glass Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K1950212 Supplier Fisher Chemical Supplier No. K195017

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Single-component reagent; Pyridine free

  • Single-component volumetric reagent for high water content analysis
  • Water equivalent: 2.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free

Specifications

Chemical Name or Material Aqualine™ Complete 2
Name Note Karl Fischer Reagent
Color Colorless
CAS 7446-09-5, 288-32-4, 111-90-0, 7553-56-2
Physical Form Liquid
Quantity 2.5 L
Molecular Formula Solution
Packaging Amber glass bottle
MDL Number 81734