missing translation for 'onlineSavingsMsg'
Learn More

AQUA™ Peptide VLVAGTSAYC[R], MilliporeSigma™ Supelco™

Catalog No. 111004123
Change view
Click to view available options
Quantity:
100 pmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004123 100 pmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004123 Supplier Merck Emd Millipore Supplier No. AQ0227100PMOL

Specifications

Product Line AQUA
Shipping Condition Wet Ice
Linear Formula C4313C6H82N1015N4O15 S
Type Standardization Solution
Quantity 100 pmol
Formula Weight 1149.25

This method was developed by Dr. Steve Gygi and colleagues at Harvard Medical School. Limited use of this method is permitted under a licensing arrangement with Harvard Medical School.