missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 FR Sleeved Apron
A single layer garment that provides triple hazard protection from liquid-chemical splash, flash fire and electric arc
$458.29 - $556.17
Specifications
| Material | Tychem |
|---|---|
| Closure Type | Waist Ties |
| Cuff Style | Elastic |
| Seam Type | Taped |
| Catalog Number | Mfr. No. | Color | Size | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Size | Price | Quantity | ||||
|
19-122-056
|
DuPont™
TP275TORMD000200 |
Orange | Medium |
Case of 2 Each for $458.29
|
|
||||
|
19-122-060
|
DuPont™
TP275TOR3X000200 |
Orange | 3X-Large |
Case of 2 Each for $514.95
|
|
||||
|
19-122-061
|
DuPont™
TP275TOR4X000200 |
Orange | 4X-Large |
Case of 2 Each for $556.17
|
|
||||
Description
- Tychem™ Thermopro for triple hazard protection
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
- Provides frontal protection
- For flash fire and electric arc hazards, partial body protective Tychem™ ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.
- Sewn with Nomex™ thread
- Two buckle closure system
- Open back for wearer comfort.
- Ideal for use on top of an existing flame resistant (FR) garment where extra chemical protection is required
- Gray color for discretionary needs
- Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Specifications
| Tychem | |
| Waist Ties | |
| Elastic | |
| Taped |