Learn More
DuPont™ Tychem™ 6000 FR Sleeved Apron
Catalog No. 19122056
Change view
Click to view available options
Color:
Orange
Size:
Medium
3X-Large
4X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19122056 Orange Medium
19122060 Orange 3X-Large
19122061 Orange 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19122056 Supplier DuPont™ Supplier No. TP275TORMD000200
A single layer garment that provides triple hazard protection from liquid-chemical splash, flash fire and electric arc
  • Tychem™ Thermopro for triple hazard protection
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Provides frontal protection
  • For flash fire and electric arc hazards, partial body protective Tychem™ ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.
  • Sewn with Nomex™ thread
  • Two buckle closure system
  • Open back for wearer comfort.
  • Ideal for use on top of an existing flame resistant (FR) garment where extra chemical protection is required
  • Gray color for discretionary needs
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Specifications
Size Medium
Material Tychem
Color Orange
Closure Type Waist Ties
Seam Type Taped