missing translation for 'onlineSavingsMsg'
Learn More

Apramycin sulfate

Apramycin sulfate, CAS # 65710-07-8, is the salt of apramycin, the bacteriostatic aminoglycoside antibiotic. | CAS: 65710-07-8 | C21H43N5O15S | 637.66 g/mol

Supplier:  Thermo Scientific Chemicals J6661609

 View more versions of this product

Catalog No. AAJ6661609


Only null left
Explore available promotions

Description

Description

Apramycin sulfate binds to the deep groove of RNA and effectively inhibits ribosomal translocation prohibiting protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Apramycin is a 2-deoxysteptamine derivative and an aminoglycoside originally isolated from Streptomyces tenebrarius
• Apramycin sulfate is comprised of a bicyclic sugar moiety and a monosubstituted deoxystreptamine attached to a sulfate group

Application

• Apramycin sulfate exhibits antibiotic activity against gram-negative bacteria
• It is suitable for laboratory use against Escherichia coli, Klebsiella pneumoniae, and Pseudomonas aeruginosa
• It is also used for laboratory study of infectious disease

Specifications

Specifications

Apramycin Sulfate
Powder
168°C
10 g
C21H43N5O15S
823°C
Soluble in water at 25mg/ml
OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O
637.66
637.65
Apramycin sulfate
65710-07-8
Yellow
>110°C (230°F)
MFCD06200257
Nebramycin II
WGLYHYWDYPSNPF-RQFIXDHTSA-N
(2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid
3081544

Chemical Identifiers

65710-07-8
637.66
WGLYHYWDYPSNPF-RQFIXDHTSA-N
3081544
OS(O)(=O)=O.CN[C@H]1[C@@H](O)[C@H]2O[C@H](O[C@@H]3[C@@H](N)C[C@@H](N)[C@H](O)[C@H]3O)[C@H](N)C[C@@H]2O[C@@H]1O[C@H]1O[C@H](CO)[C@@H](N)[C@H](O)[C@H]1O
C21H43N5O15S
MFCD06200257
Nebramycin II
(2R,3R,4S,5S,6S)-2-{[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-{[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy}-4-hydroxy-3-(methylamino)-octahydropyrano[3,2-b]pyran-2-yl]oxy}-5-amino-6-(hydroxymethyl)oxane-3,4-diol; sulfuric acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H360-H315-H319; May damage fertility or the unborn child. Causes skin irritation. Causes serious eye irritation.

P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H360

EINECSNumber : 265-890-7

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only