Learn More
RapidHIT™ ID ACE GlobalFiler™ Express 50 Sample Kit
Catalog No. A41831
Change view
Click to view available options
Quantity:
1 kit
Catalog No. Quantity
A41831 1 kit
1 options
Catalog No. A41831 Supplier Thermo Fisher Scientific Supplier No. A41831
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Applied Biosystems RapidHIT ID ACE GlobalFiler Express 50 Sample Kit provides 50 self-contained, easy-to-use sample cartridges that contain all necessary reagents for sample preparation, including GlobalFiler Express chemisty, for sample preparation.
NOTE: For use with the Applied Biosystems-branded RapidHIT ID System only (Cat. No. A41810).
The Applied Biosystems RapidHIT ID ACE GlobalFiler Express 50 Sample Kit provides 50 self-contained, easy-to-use sample cartridges that contain all necessary reagents for sample preparation, including GlobalFiler Express chemisty, for sample preparation. With less than one minute of hands-on time, users simply insert the sample (via swab) into the sample cartridge, insert the sample cartridge into the instrument, and in 90 minutes receive the lab-quality forensic DNA profile.
Features of the kit include:
  • 1-minute hands-on-time workflow provided by self-contained, single-use sample cartridges
  • Easy consumables information tracking and auto-selection of instrument run parameters through integrated radio frequency identification (RFID) tagging in cartridge
The kit includes:
  • 50 RapidHIT ID ACE GlobalFiler Express sample cartridges
  • 2 positive control RapidHIT ID ACE GlobalFiler Express sample cartridges
  • 2 negative control RapidHIT ID ACE GlobalFiler Express sample cartridges
Specifications
Content And Storage • 50 RapidHIT ID ACE GlobalFiler Express sample cartridges
• 2 positive control RapidHIT ID ACE GlobalFiler Express sample cartridges
• 2 negative control RapidHIT ID ACE GlobalFiler Express sample cartridges
Product Line RapidHIT
Quantity 1 kit