missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ Adhesive Film Applicator

Catalog No. 4333183
Encompass
Change view
Click to view available options
Quantity:
5 applicators
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4333183 5 applicators
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4333183 Supplier Applied Biosystems™ Supplier No. 4333183
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Five Adhesive Seal Applicators

Use to form a secure seal between a microplate and an adhesive film

Form a Tight Seal

  • The MicroAmp Adhesive Film Applicator enables you to ergonomically push an adhesive film down upon a microplate to form a tight seal and minimize air bubbles
  • This tight seal will prevent well-to-well contamination and sample loss
Quick and Easy
  • Use the MicroAmp Adhesive Film Applicator to speed the process of sealing multiple microplates
  • This tool is designed according to the microplate dimensions so you can quickly seal a microplate
  • The rugged proprietary plastic endures constant use for large screening applications

See user's manual or package insert for limited label license and trademark information.

PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)

Order Info

Shipping Condition: Room Temperature

Specifications

Product Line MicroAmp
Certifications/Compliance Certified DNA/RNase Free
Color Natural
Includes Includes: 5 Adhesive Seal Applicators
Material Polyethylene
No. per Pack Pack of 5
Storage Requirements Room Temperature
Product Type Adhesive Film Applicator
Quantity 5 applicators
How do I correctly apply MicroAmp Optical Adhesive Covers to an optical 96- or 384-well plate?

Please follow the instructions below for the proper application of MicroAmp Optical Adhesive Covers (Cat. Nos. 4311971, 4313663 and 4360954).

The MicroAmp Optical Adhesive Cover uses a pressure-sensitive adhesive backing to adhere the cover to the optical 96- or 384-well plate. It is imperative that you use enough force to activate the pressure-sensitive adhesive to prevent evaporation from the wells.
Please note: Users should not expect the Optical Adhesive Cover to be very sticky to the touch on the side that adheres to the optical plate.

The correct way to apply the cover is as follows:

1. Apply firm pressure when using the Adhesive Seal Applicator (Cat. No. 4333183) to press the cover onto the plate.

2. Run the applicator firmly from left to right several times. Run the applicator up and down several times.

3. Run the edge of the applicator around the border (following the red line outside the wells) of the cover.

4. Repeat the left to right and up and down motions to assure a good final seal.

5. Inspect each well to make sure that each well edges are imprinted on the optical adhesive cover.
Please Note: Using the Applicator's edge to seal between the wells is NOT recommended. It may actually pull the seal off the wells.