missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ OpenArray™ Plate Compression Pad for Dual Flat Block GeneAmp™ PCR System 9700

Catalog No. 4426825
Encompass
Change view
Click to view available options
Quantity:
1 pad
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4426825 1 pad
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4426825 Supplier Applied Biosystems™ Supplier No. 4426825

Properly and evenly positions up to eight TaqMan™ OpenArray™ Genotyping Plates for PCR amplification

It is intended for use with the Dual Flat Block GeneAmp™ PCR System 9700 System and OpenArray™ Plate Adaptor for Dual Flat Block GeneAmp™ PCR System 9700.

  • Platform: GeneAmp™ 9700
  • Product Size: 1 pad
  • Accessory Type: Compression Pad
  • Plate Compatibility: OpenArray™ Plates
  • Shipping Condition: Room Temperature

Specifications

Content And Storage Required compression pad for using OpenArray™ plate holder with the GeneAmp 9700 Dual flat block PCR system
For Use With (Equipment) GeneAmp 9700
Product Line GeneAmp, OpenArray
Product Type Plate Compression Pad
For Use With (Application) PCR
Shipping Condition Room Temperature
Quantity 1 pad
Plate Compatibility OpenArray™ Plates

For Research Use Only. Not for use in diagnostic procedures.