missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ OpenArray™ Plate Adaptor for Dual Flat Block GeneAmp™ PCR System 9700

Catalog No. 4426824
Encompass
Change view
Click to view available options
Quantity:
1 adaptor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4426824 1 adaptor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4426824 Supplier Applied Biosystems™ Supplier No. 4426824

Includes

Required adaptor for using OpenArray™ plate holder with the GeneAmp 9700 Dual flat block PCR system

Serves to secure up to eight TaqMan™ OpenArray™ Genotyping Plates in place for PCR amplification

  • Use the Plate Adaptor with the Dual Flat Block GeneAmp™ PCR System 9700 System and OpenArray Plate Compression Pad for Dual Flat Block GeneAmp PCR System 9700
  • Accessory Type: Adaptor

Gene Expression Analysis & Genotyping, Genotyping & Genomic Profiling, PCR, PCR & Real-Time PCR

Specifications

Content And Storage Required adaptor for using OpenArray™ plate holder with the GeneAmp 9700 Dual flat block PCR system
For Use With (Equipment) GeneAmp 9700
Product Line GeneAmp, OpenArray
Product Type Thermal Cycler Accessories
For Use With (Application) PCR
Shipping Condition Room Temperature
Quantity 1 adaptor
Plate Compatibility OpenArray™ Plates
I have an Applied Biosystems GeneAmp PCR System 9700. What additional accessories do I need to perform off-line cycling for Applied Biosystems QuantStudio OpenArray plates?

You need an Applied Biosystems OpenArray Plate Adaptor (Part Number 4426824). You do not need a compression pad.

For Research Use Only. Not for use in diagnostic procedures.