missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APPL1 Recombinant Rabbit Monoclonal Antibody (23GB1570)
GREENER_CHOICE

Catalog No. MA553668
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553668 100 μL
MA553669 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553668 Supplier Invitrogen™ Supplier No. MA553668
Only null left

Rabbit Recombinant Monoclonal Antibody

APPL1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The protein encoded by this gene has been shown to be involved in the regulation of cell proliferation, and in the crosstalk between the adiponectin signaling and insulin signaling pathways. The encoded protein binds many other proteins, including RAB5A, DCC, AKT2, PIK3CA, adiponectin receptors, and proteins of the NuRD/MeCP1 complex. This protein is found associated with endosomal membranes, but can be released by EGF and translocated to the nucleus.

Specifications

Antigen APPL1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1570
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene APPL1
Gene Accession No. Q8K3H0, Q9UKG1
Gene Alias 2900057D21Rik; 7330406P05Rik; Adapter protein containing PH domain, PTB domain and leucine zipper motif 1; adaptor protein APPL like; adaptor protein containing pH domain, PTB domain and leucine zipper motif 1; adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1; adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1; AI585782; AKT2 interactor; APPL; Appl1; AW209077; BB022931; C88264; DCC-interacting protein 13-alpha; DIP13; DIP13 alpha; DIP13A; DIP13alpha; dip13-alpha; Kiaa1428; MODY14; RGD1309388; signaling adaptor protein DIP13alpha
Gene Symbols APPL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human APPL (400-450AA).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26060, 290537, 72993
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less