missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apolipoprotein L5 Polyclonal Antibody

Description
Antibody detects endogenous levels of total APOL5.
This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles.
Specifications
Specifications
| Antigen | Apolipoprotein L5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | APOL5 |
| Gene Accession No. | Q9BWW9 |
| Gene Alias | APOL5; apolipoprotein L, 5; Apolipoprotein L5; apolipoprotein L-V; APOLV; APOL-V; OTTHUMP00000028773 |
| Gene Symbols | APOL5 |
| Show More |