accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Facility Safety and Maintenance
Cleaning Supplies and Equipment
Cleaners and Disinfectants
Laboratory Surfactants and Wetting Agents
1783751GM
Structure Search
Print
MilliporeSigma™ APO-10, Calbiochem™,
Catalog No.
1783751GM
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
1783751GM
1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
1783751GM
Supplier
MilliporeSigma™
Supplier No.
1783751GM
Sign In
or
Register
to check your price and availability.
Quantity
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Chemical Identifiers
CAS
2190-95-6
Molecular Formula
C12H27OP
Molecular Weight (g/mol)
218.321
InChI Key
GSVLCKASFMVUSW-UHFFFAOYSA-N
Synonym
Dimethyldecylphosphine Oxide
PubChem CID
3492020
IUPAC Name
1-dimethylphosphoryldecane
SMILES
CCCCCCCCCCP(=O)(C)C
Specifications
Color
White
Quantity
1 g
Packaging
Glass Bottle
Product Suggestions
Documents