accessibility menu, dialog, popup

UserName

H3CPCH3CH3O

CAS RN 2190-95-6

IUPAC Name: 1-(dimethylphosphoryl)decane
Synonyms: decyl(dimethyl)phosphine oxide
Molecular Weight (g/mol): 218.32
Molecular Formula: C12H27OP
InChi Key: GSVLCKASFMVUSW-UHFFFAOYSA-N
SMILES: CCCCCCCCCCP(C)(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results