missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APLP2 Recombinant Rabbit Monoclonal Antibody (23GB1595)

Catalog No. MA553665
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553665 20 μL
MA553664 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553665 Supplier Invitrogen™ Supplier No. MA553665
Only null left

Rabbit Recombinant Monoclonal Antibody

APLP2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

APLP2 May play a role in the regulation of hemostasis. The soluble form may have inhibitory properties towards coagulation factors. May interact with cellular G-protein signaling pathways. May bind to the DNA 5'-GTCACATG-3' (CDEI box).

Specifications

Antigen APLP2
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1595
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene APLP2
Gene Accession No. Q06481
Gene Alias AI790698; amyloid beta (A4) precursor-like protein 2; amyloid beta (A4) precursor-like protein 2-like; amyloid beta precursor like protein 2; amyloid precursor protein homolog HSD-2; Amyloid protein homolog; Amyloid protein precursor-like protein 2; amyloid-like protein 2; APLP2; APLP-2; APPH; APPL2; CDEBP; CDEI box-binding protein; sperm membrane protein (YWK-II); sperm membrane protein YWK-II; testicular tissue protein Li 23; WALPLP2
Gene Symbols APLP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human APLP2 (221-280AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 334
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less