missing translation for 'onlineSavingsMsg'
Learn More

Apigenin-7-Glucoside Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111001981
Change view
Click to view available options
Quantity:
75 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001981 75 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001981 Supplier Merck Emd Millipore Supplier No. PHR219975MG

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 578-74-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C21H20O10
MDL Number MFCD00016787
Synonym Apigenin 7-glucoside; 4′,5,7-Trihydroxyflavone 7-glucoside; Apigetrin; Cosmosiin
Type Certified Reference Material
Quantity 75 mg
Formula Weight 432.38
Grade Certified Reference Material