missing translation for 'onlineSavingsMsg'
Learn More

Apigenin 7-glucoside, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014733
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014733 25 mg
111014732 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014733 Supplier Merck Emd Millipore Supplier No. 4469225MGF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Apigenin 7-glucoside is a flavone belonging to the class of polyphenols and is commonly found in Thymus species. It shows a broad spectrum of pharmacological properties such as antispasmodic, bactericidal, expectorant, antihelmintic, adstringent, antitussive and carminative agent.

Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 578-74-5
Physical Form Neat
Linear Formula C21H20O10
MDL Number MFCD00016787
Synonym 4 ′,5,7-Trihydroxyflavone 7-glucoside; Apigetrin; Cosmosiin
Type Analytical Standard
Quantity 25 mg
Formula Weight 432.38
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Show More Show Less