Learn More
Thermo Scientific Chemicals Aphidicolin
A DNA polymerase inhibitor, blocks the cell cycle at the early S-phase | CAS: 38966-21-1 | C20H34O4, C20H34O4 | 338.49 g/mol
Supplier: Thermo Scientific Chemicals J60236LB0
| Quantity | 5 mg |
|---|
Description
A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 38966-21-1 | |
| C20H34O4 | |
| APC |
Specifications
| 5 mg | |
| 2°C to 8°C | |
| 38966-21-1 | |
| 338.48 | |
| 224°C | |
| Aphidicolin | |
| 4689958 | |
| C[C@@]1(CO)[C@H](O)CC[C@@]2(C)[C@H]1CC[C@H]1C[C@@H]3C[C@]21CC[C@]3(O)CO | |
| 338.49 |
| White | |
| A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase | |
| C20H34O4 | |
| APC | |
| Soluble in DMSO or methanol. Insoluble in water. Store solutions at -20° | |
| Cyrstalline powder | |
| NOFOAYPPHIUXJR-APNQCZIXSA-N | |
| (1S,2S,5R,6R,7R,10S,12R,13R)-6,13-bis(hydroxymethyl)-2,6-dimethyltetracyclo[10.3.1.0¹,¹⁰.0²,⁷]hexadecane-5,13-diol |
Safety and Handling
Recommended Storage : Keep cold