missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ APH1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113140 Supplier Invitrogen™ Supplier No. PA5113140
Only null left

Rabbit Polyclonal Antibody

APH1B Polyclonal Antibody for Western Blot, ELISA

APH1B gene ontology annotations related to this gene include Notch receptor processing; Notch signaling pathway; positive regulation of catalytic activity; protein processing.

Specifications

Antigen APH1B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene APH1B
Gene Accession No. Q8WW43
Gene Alias 2310057K14Rik; anterior pharynx defective 1 homolog B; anterior pharynx defective 1b homolog; anterior pharynx defective 1b homolog (C. elegans); anterior pharynx defective 1c homolog; aph-1 homolog B, gamma secretase subunit; aph-1 homolog B, gamma-secretase subunit; Aph1b; APH-1b; APH-1B'; APH1B gamma secretase subunit; APH1B homolog; aph-1beta; Aph1c; Gamma-secretase subunit APH-1B; LOW QUALITY PROTEIN: gamma-secretase subunit APH-1B; Presenilin-stabilization factor-like; PRO1328; PSFL; TAAV688; UNQ688/PRO1328
Gene Symbols APH1B
Host Species Rabbit
Immunogen Peptide sequence from Human Gamma-secretase subunit APH-1B protein (93-110AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83464
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less