missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANAPC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584731
Description
Immunogen sequence: REEVHTMLRG VLFFSTPRTF QEMIQRLYGC FLRVYMQSKR KGEGGTDPEL EGELDSRYAR RRYYRLLQSP LCAGCSSDKQ QCWCR Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 95%.
A large protein complex, termed the anaphase-promoting complex (APC), or the cyclosome, promotes metaphase-anaphase transition by ubiquitinating its specific substrates such as mitotic cyclins and anaphase inhibitor, which are subsequently degraded by the 26S proteasome. Biochemical studies have shown that the vertebrate APC contains eight subunits.Specifications
| ANAPC2 | |
| Polyclonal | |
| Unconjugated | |
| ANAPC2 | |
| 9230107K09Rik; AL024279; Anapc2; anaphase promoting complex subunit 2; anaphase-promoting complex subunit 2; Apc2; cyclosome subunit 2; Emi4; expressed during mesenchymal induction 4; Imi4; induced during mesenchymal induction 4; KIAA1406; mKIAA1406 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29882 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UJX6 | |
| ANAPC2 | |
| Recombinant protein corresponding to Human ANAPC2. Recombinant protein control fragment (Product #RP-105397). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |