missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apaf-1 Antibody [5E11] , ProSci
Specifications
Specifications
| Antigen | Apaf-1 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 5E11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Apaf1 antibody can be used for detection of Apaf1 by immunohistochemistry at 0.5 mg/mL. Apaf1 has not been confirmed for use in Western Blot. Antibody validated: Immunohistochemistry in human samples. All other applications and species not yet tested. |
| Formulation | Apaf-1 Monoclonal Antibody is supplied in PBS containing 0.02% sodium azide. |
| Gene Accession No. | AAC51678 |
| Gene Alias | Apaf-1 Antibody [5E11] : CED4, APAF-1 |
| Show More |