missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AP2 alpha Monoclonal Antibody (K92029_15F12), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555880
Description
Sequence of this protein is as follows: EDLLHGPHAL SSGLGDLSIH SLPHAIEEVP HVEDPGINIP DQTVIKKGPV SLSKSNSNAV SAIPINKDNL FGGVVNPNEV FCSVPGRLSL LSSTSKYKVT VAEVQRRLSP PECLNASLLG GVLRRAKSKN GGRSLREKLD KIGLNLPAGR RKAANVTLLT SLVEGEAVHL ARDFGYVCET EFPAKAVAEF LNRQHSDPNE QVTRKNMLLA TKQICKEFTD LLAQDRSPLG NSRPNPILEP GIQSCLTHFN LISHGFGSPA VCAAVTALQN YLTEALKAMD KMYLSNNPNS HTDNNAKSSD
TFAP2A is a sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5'-GCCNNNGGC-3' and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limbs and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC. TFAP2A is the only AP-2 protein required for early morphogenesis of the lens vesicle.Chemical Identifiers
| AP2 alpha | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| activating enhancer-binding protein 2 alpha; activating enhancer-binding protein 2-alpha; Activator protein 2; activator protein 2 alpha variant 4; activator protein 2 alpha variant 6; activator protein 2 alpha variant 7; AP2; AP-2; Ap-2 (a); AP-2 transcription factor; AP2alpha; AP-2alpha; AP2-alpha; Ap2tf; BOFS; ERF-1; FLJ51761; OTTHUMP00000214240; OTTHUMP00000214243; Tcfap2a; TFAP2; TFAP2A; transcription factor AP2; transcription factor AP-2 alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); transcription factor AP-2, alpha; transcription factor AP-2-alpha; transcription factor AP-2-alpha-like; transcription factor; AP-2 | |
| Recombinant human TFAP2A (immunogen range is 131-430). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| TFAP2A |
| K92029_15F12 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P05549 | |
| 7020 | |
| IgG2a | |
| Protein A | |
| Antibody |
Specifications
| AP2 alpha | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P05549 | |
| TFAP2A | |
| Recombinant human TFAP2A (immunogen range is 131-430). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K92029_15F12 | |
| Unconjugated | |
| TFAP2A | |
| activating enhancer-binding protein 2 alpha; activating enhancer-binding protein 2-alpha; Activator protein 2; activator protein 2 alpha variant 4; activator protein 2 alpha variant 6; activator protein 2 alpha variant 7; AP2; AP-2; Ap-2 (a); AP-2 transcription factor; AP2alpha; AP-2alpha; AP2-alpha; Ap2tf; BOFS; ERF-1; FLJ51761; OTTHUMP00000214240; OTTHUMP00000214243; Tcfap2a; TFAP2; TFAP2A; transcription factor AP2; transcription factor AP-2 alpha; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); transcription factor AP-2, alpha; transcription factor AP-2-alpha; transcription factor AP-2-alpha-like; transcription factor; AP-2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 7020 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |