missing translation for 'onlineSavingsMsg'
Learn More

AOCS Low Erucic Rapeseed Oil, MilliporeSigma™ Supelco™

Catalog No. 111002703
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002703 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002703 Supplier Merck Emd Millipore Supplier No. O77561AMP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Our quantitative animal and vegetable reference mixes conform to the requirements of the American Oil Chemists' Society (AOCS) method Ce 1-62. The composition of each mix is similar to the fatty acid distribution of certain oils listed under each product. Please note that the AOCS mixtures sold under the Supelco brand are named RM-1, RM-2, etc.

Specifications

Packaging Ampule
CAS 124-10-7
Shipping Condition Dry Ice
Physical Form Neat
Synonym AOCS Reference Mixtures; F.A.M.E. Mix Rapeseed Oil; Rapeseed oil mixture; Oil Reference Standard, AOCS
Type Analytical Standard
Quantity 100 mg
Grade Analytical Standard