missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AOAH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103566
Description
Antibody detects endogenous levels of total AOAH.
Removes the secondary (acyloxyacyl-linked) fatty acyl chains from the lipid A region of bacterial lipopolysaccharides.Chemical Identifiers
| AOAH | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| AOAH | |
| 4930433E13Rik; Acyloxyacyl hydrolase; acyloxyacyl hydrolase (neutrophil); Acyloxyacyl hydrolase large subunit; Acyloxyacyl hydrolase small subunit; AOAH; RGD1560514 | |
| AOAH | |
| Primary | |
| Affinity Chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35298, P28039 | |
| 27052, 313, 498757 | |
| A synthesized peptide derived from human AOAH(Accession P28039), corresponding to amino acid residues S302-E352. | |
| Antibody |
Specifications
| AOAH | |
| Polyclonal | |
| Unconjugated | |
| AOAH | |
| 4930433E13Rik; Acyloxyacyl hydrolase; acyloxyacyl hydrolase (neutrophil); Acyloxyacyl hydrolase large subunit; Acyloxyacyl hydrolase small subunit; AOAH; RGD1560514 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27052, 313, 498757 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35298, P28039 | |
| AOAH | |
| A synthesized peptide derived from human AOAH(Accession P28039), corresponding to amino acid residues S302-E352. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |