missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anza™ 5-Pack Starter Kit

Catalog No. IVGN3004
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IVGN3004 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IVGN3004 Supplier Invitrogen™ Supplier No. IVGN3004

Includes

Anza™ 11 EcoRI, Anza™ 5 BamHI, Anza™ 1 NotI, Anza™ 16 HindIII, Anza™ 12 XbaI, Anza™ T4 DNA Ligase Master Mix, Anza™ Alkaline Phosphatase, Anza™ 10X Buffer, Anza™ 10X Red Buffer,

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • One buffer for all restriction enzymes
  • One digestion protocol for all DNA substrates
  • Complete digestion in 15 min
  • Includes 5 common restriction enzymes
  • Complete with Anza T4 DNA Ligase Master Mix and Anza Alkaline Phosphatase

Certifications

CoA

Specifications

Content And Storage • 20 μL each of Anza 11 EcoRI, Anza 5 BamHI, Anza 1 NotI, Anza 16 HindIII, Anza 12 XbaI
• 100 μL Anza T4 DNA Ligase Master Mix
• 20 μL Anza Alkaline Phosphatase
• 500 μL Anza Buffer (10X)
• 500 μL Anza Red Buffer (10X)

Store at -5 to -30°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Methylation Sensitivity Methylation Sensitivities Varies by Enzyme
Enzyme HindIII, NotI, Alkaline Phosphatase, XbaI, T4 DNA Ligase, BamHI, EcoRI
Compatible Buffer Anza™ 10X Red Buffer, Anza™ 10X Buffer
Type IIS RE No
Quantity 1 kit
Product Type Starter Kit
Research Category Traditional Cloning
Product Line Anza

For Research Use Only. Not for use in diagnostic procedures.