missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Anza™ 5 BamHI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Supplier: Invitrogen™ IVGN0058
Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity
Certifications
CoA
Chemical Identifiers
| Approved for shipment on Wet or Dry Ice | |
| BamH I | |
| Yes | |
| No | |
| Traditional Cloning |
| Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive | |
| Anza™ 10X Red Buffer, Anza™ 10X Buffer | |
| 37°C | |
| Restriction Enzyme Cloning System | |
| Anza |
Specifications
| 2.4 mL Anza 5 BamHI 5 mL Anza Buffer (10X) 5 mL Anza Red Buffer (10X) Store at -20°C. |
|
| Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive | |
| Anza™ 10X Red Buffer, Anza™ 10X Buffer | |
| 37°C | |
| 24,000 units | |
| Traditional Cloning |
| Approved for shipment on Wet or Dry Ice | |
| BamH I | |
| Yes | |
| No | |
| Restriction Enzyme Cloning System | |
| Anza |
For Research Use Only. Not for use in diagnostic procedures.