missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anza™ Buffer Set

Catalog No. IVGN2008
Encompass
Change view
Click to view available options
Quantity:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IVGN2008 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IVGN2008 Supplier Invitrogen™ Supplier No. IVGN2008

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Anza™ 10X Buffer, Anza™ 10X Red Buffer

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • One buffer for all restriction enzymes
  • One digestion protocol for all DNA types
  • Complete digestion in 15 min
  • Overnight digestion without star activity

Certifications

CoA

Specifications

Chemical Name or Material Buffer Set
For Use With (Application) Traditional Cloning
Product Line Anza
Quantity 1 set
Includes 2 mL Anza Buffer (10X)
2 mL Anza Red Buffer (10X)

Store at -5 to -30°C.
Is BSA present in the Anza Buffers?

As an added convenience, BSA is included in the Anza 10X Buffers and Anza 10X Red Buffers. This eliminates the need to add BSA in a separate step.

Can I store the Anza Buffers at 4 degrees C?

The Anza Buffers can be stored at 4 degrees C for at least 6 months. To maintain effectiveness through the expiration date, we recommend longer term storage of both the Anza 10X Buffer and Anza 10X Red Buffer at -20 degrees C.

For Research Use Only. Not for use in diagnostic procedures.