missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Anza™ 10 DpnI
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Specifications
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
|---|---|
| Methylation Sensitivity | Not CpG Methylation-Sensitive |
| Enzyme | Dpn I |
| Compatible Buffer | Anza™ 10X Red Buffer, Anza™ 10X Buffer |
| Sensitive to Heat Inactivation | Yes |
Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity
Certifications
CoA
Specifications
| Approved for shipment on Wet or Dry Ice | |
| Dpn I | |
| Yes | |
| No | |
| Traditional Cloning |
| Not CpG Methylation-Sensitive | |
| Anza™ 10X Red Buffer, Anza™ 10X Buffer | |
| 37°C | |
| Restriction Enzyme Cloning System | |
| Anza |
For Research Use Only. Not for use in diagnostic procedures.