missing translation for 'onlineSavingsMsg'
Learn More

Antipain, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215284325

 View more versions of this product

Catalog No. ICN15284325

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Antipain
Yellow
MFCD00135959
antipain, antipain hydrochloride dihydrate, antipain, >50000 u/mg, s-1-carboxy-2-phenylethyl carbamoyl-l-arginyl-l-valyl-argininal, 2s-2-1s-4-carbamimidamido-1-1s-1-5-carbamimidamido-1-oxopentan-2-yl carbamoyl-2-methylpropyl carbamoyl butyl carbamoyl amino-3-phenylpropanoic acid
CC(C)[C@H](NC(=O)[C@H](CCCN=C(N)N)NC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O)C(=O)N[C@@H](CCCN=C(N)N)C=O
604.71
604.7
37691-11-5
C27H44N10O6
25 mg
SDNYTAYICBFYFH-TUFLPTIASA-N
2-[[(2R)-5-(diaminomethylideneamino)-1-[[1-[[5-(diaminomethylideneamino)-1-oxopentan-2-yl]amino]-3-methyl-1-oxobutan-2-yl]amino]-1-oxopentan-2-yl]carbamoylamino]-3-phenylpropanoic acid
133126507
Powder

Chemical Identifiers

37691-11-5
604.71
SDNYTAYICBFYFH-TUFLPTIASA-N
133126507
CC(C)[C@H](NC(=O)[C@H](CCCN=C(N)N)NC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O)C(=O)N[C@@H](CCCN=C(N)N)C=O
C27H44N10O6
MFCD00135959
antipain, antipain hydrochloride dihydrate, antipain, >50000 u/mg, s-1-carboxy-2-phenylethyl carbamoyl-l-arginyl-l-valyl-argininal, 2s-2-1s-4-carbamimidamido-1-1s-1-5-carbamimidamido-1-oxopentan-2-yl carbamoyl-2-methylpropyl carbamoyl butyl carbamoyl amino-3-phenylpropanoic acid
2-[[(2R)-5-(diaminomethylideneamino)-1-[[1-[[5-(diaminomethylideneamino)-1-oxopentan-2-yl]amino]-3-methyl-1-oxobutan-2-yl]amino]-1-oxopentan-2-yl]carbamoylamino]-3-phenylpropanoic acid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions