missing translation for 'onlineSavingsMsg'
Learn More

Antipain dihydrochloride

Catalog No. AAJ63680LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63680LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63680LB0 Supplier Thermo Scientific Chemicals Supplier No. J63680LB0
Only null left

CAS: 37682-72-7 | C27H46Cl2N10O6 | 677.63 g/mol

Antipain inhibits trypsin, papain, and catherpsins A and B (a reversible inhibitor of cysteine and serine proteases). It is used to evaluate the role of proteases in cell transformations. It is used to help identify new proteases.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Antipain inhibits trypsin, papain, and catherpsins A and B (a reversible inhibitor of cysteine and serine proteases). It is used to evaluate the role of proteases in cell transformations. It is used to help identify new proteases.

Chemical Identifiers

CAS 37682-72-7
Molecular Formula C27H46Cl2N10O6
Molecular Weight (g/mol) 677.63
MDL Number MFCD00135957
InChI Key GXQPAKIVOCLCHD-BXLPLHKWSA-N
Synonym antipain dihydrochloride
PubChem CID 24847649
IUPAC Name 2-[[5-(diaminomethylideneamino)-1-[[1-[[5-(diaminomethylideneamino)-1-oxopentan-2-yl]amino]-3-methyl-1-oxobutan-2-yl]amino]-1-oxopentan-2-yl]carbamoylamino]-3-phenylpropanoic acid;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].CC(C)[C@H](NC(CCCN=C(N)N)C=O)C(=O)NC(=O)[C@H](CCCN=C(N)N)NC(=O)NC(CC1=CC=CC=C1)C(O)=O

Specifications

Color White to Yellow
Quantity 5 mg
Solubility Information 0.01M in water
Formula Weight 677.6
Concentration or Composition (by Analyte or Components) 0.01M in water
Physical Form Powder
Chemical Name or Material Antipain dihydrochloride
RTECSNumber YV9350800
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only