accessibility menu, dialog, popup

UserName

ClClCH3H3C(S)HNNH2NH2NOONHO(S)NH2NNH2HNOHNOHOHH

CAS RN 37682-72-7

IUPAC Name: dihydrogen 2-({[(1S)-4-[(diaminomethylidene)amino]-1-{[(2S)-2-({5-[(diaminomethylidene)amino]-1-oxopentan-2-yl}amino)-3-methylbutanoyl]carbamoyl}butyl]carbamoyl}amino)-3-phenylpropanoic acid dichloride
Synonyms: dihydrogen 2-({[(1S)-4-[(diaminomethylidene)amino]-1-{[(2S)-2-({5-[(diaminomethylidene)amino]-1-oxopentan-2-yl}amino)-3-methylbutanoyl]carbamoyl}butyl]carbamoyl}amino)-3-phenylpropanoic acid dichloride
Molecular Weight (g/mol): 677.63
Molecular Formula: C27H46Cl2N10O6
InChi Key: GXQPAKIVOCLCHD-BXLPLHKWSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CC(C)[C@H](NC(CCCN=C(N)N)C=O)C(=O)NC(=O)[C@H](CCCN=C(N)N)NC(=O)NC(CC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results