missing translation for 'onlineSavingsMsg'
Learn More

Antimony Trioxide, 99%, Spectrum™ Chemical

Catalog No. 18608913
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608913 2.5 kg Amber Glass Bottle
18608909 125 g Amber Glass Bottle
18608911 500 g Amber Glass Bottle
18608910 12 kg Poly Pail
18608912 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608913 Supplier Spectrum Chemical Mfg Cor Supplier No. A13302.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1309-64-4

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 99%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.