Learn More
Antimony tribromide, 97%, pure
CAS: 7789-61-9 | Br3Sb | 361.48 g/mol
$234.16 - $234.16
Chemical Identifiers
| CAS | 7789-61-9 |
|---|---|
| Molecular Formula | Br3Sb |
| Molecular Weight (g/mol) | 361.48 |
| InChI Key | RPJGYLSSECYURW-UHFFFAOYSA-K |
| Synonym | antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3 |
| PubChem CID | 24615 |
| IUPAC Name | tribromostibane |
| SMILES | Br[Sb](Br)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC318400500
|
Thermo Scientific Chemicals
318400500 |
50 g |
Each for $234.16
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7789-61-9 | |
| 361.48 | |
| antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3 | |
| tribromostibane |
| Br3Sb | |
| RPJGYLSSECYURW-UHFFFAOYSA-K | |
| 24615 | |
| Br[Sb](Br)Br |
Specifications
| 280.0°C | |
| 7789-61-9 | |
| 96% min. (Argentometry) | |
| SbBr3 | |
| antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3 | |
| Br[Sb](Br)Br | |
| 361.48 | |
| 361.48 | |
| Pure | |
| Antimony tribromide |
| 97.0°C | |
| 50 g | |
| Br3Sb | |
| 15,695 | |
| RPJGYLSSECYURW-UHFFFAOYSA-K | |
| tribromostibane | |
| 24615 | |
| 97% | |
| Glass bottle |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Dispose of contents/container to an approve
GHS Signal Word: Warning
EINECSNumber : 232-179-8
RUO – Research Use Only