missing translation for 'onlineSavingsMsg'
Learn More

Antimony tribromide, 97%, pure

Catalog No. ac318400500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC318400500 50 g
AC318402500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC318400500 Supplier Thermo Scientific Chemicals Supplier No. 318400500

CAS: 7789-61-9 | Br3Sb | 361.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-61-9
Molecular Formula Br3Sb
Molecular Weight (g/mol) 361.48
InChI Key RPJGYLSSECYURW-UHFFFAOYSA-K
Synonym antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
PubChem CID 24615
IUPAC Name tribromostibane
SMILES Br[Sb](Br)Br

Specifications

Boiling Point 280.0°C
Melting Point 97.0°C
Quantity 50 g
Assay Percent Range 96% min. (Argentometry)
Linear Formula SbBr3
Merck Index 15,695
Formula Weight 361.48
Percent Purity 97%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Antimony tribromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Dispose of contents/container to an approve
EINECSNumber 232-179-8

RUO – Research Use Only