missing translation for 'onlineSavingsMsg'
Learn More

Antimony shot, 6.35mm (0.25in) & down, 99% (metals basis)

Catalog No. aa4509536
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
100 g
500 g
Catalog No. Quantity
AA4509536 500 g
AA4509522 100 g
AA45095A1 1 kg
3 options

Catalog No. AA4509536

Supplier: Thermo Scientific Chemicals 04509536

Only null left

CAS: 7440-36-0 | Sb | 121.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-36-0
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
Synonym powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
PubChem CID 5354495
ChEBI CHEBI:30304
SMILES [Sb]

Specifications

Color Silver to Gray
Appearance Silver Gray shot
Quantity 500 g
Odor Odorless
Form Shot
Assay Percent Range 99% (Metals basis)
IUPAC Name antimony
Chemical Name or Material Antimony
EINECSNumber 231-146-5
RTECSNumber CC4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only