missing translation for 'onlineSavingsMsg'
Learn More

Antimony powder, -200 mesh, 99.5% (metals basis)

Catalog No. AA1009909
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1009909 10 g
AA1009922 100 g
AA1009936 500 g
AA10099A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1009909 Supplier Thermo Scientific Chemicals Supplier No. 01009909

CAS: 7440-36-0 | Sb | 121.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-36-0
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
Synonym powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
PubChem CID 5354495
ChEBI CHEBI:30304
SMILES [Sb]

Specifications

Color Gray
Quantity 10 g
Mesh Size −200 Mesh
Odor Odorless
Form Powder
UN Number UN2871
IUPAC Name antimony
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Antimony powder
Hazard Category H335-H351-H373
Hazard Statement GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P271-P281-P304+P340-P308+P313-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY POWDER
EINECSNumber 231-146-5
RTECSNumber CC4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only