missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) sulfide, 99.9% (metals basis)

Vacuum deposition | CAS: 1345-04-6 | S3Sb2 | 339.70 g/mol

Supplier:  Thermo Scientific Chemicals 04037118

 View more versions of this product

Catalog No. AA4037118


Only null left
Explore available promotions

Description

Description

Antimony Trisulfide is used in the production of pyrotechnics and as a yellow pigment for glass and porcelain. In Vacuum deposition. Used in preparation of Vermilion or yellow pigment, for antimony salts, percussion caps, camouflage paints, and in ruby glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Antimony(III) sulfide
1345-04-6
50 g
S3Sb2
UN1549
Insoluble in water. Soluble in concentrated acids.
[S--].[S--].[S--].[Sb+3].[Sb+3]
339.70
99.9%
550°C
Pieces
(metals basis)
MFCD00011217
14,713
NVWBARWTDVQPJD-UHFFFAOYSA-N
diantimony(3+) trisulfanediide
339.68

Chemical Identifiers

1345-04-6
339.70
NVWBARWTDVQPJD-UHFFFAOYSA-N
[S--].[S--].[S--].[Sb+3].[Sb+3]
S3Sb2
MFCD00011217
diantimony(3+) trisulfanediide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H312-H315-H319-H335-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.

EINECSNumber : 215-713-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only