missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony(III) sulfide, 99.9% (metals basis)
Description
Antimony Trisulfide is used in the production of pyrotechnics and as a yellow pigment for glass and porcelain. In Vacuum deposition. Used in preparation of Vermilion or yellow pigment, for antimony salts, percussion caps, camouflage paints, and in ruby glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 550°C |
| Physical Form | Pieces |
| Quantity | 10 g |
| UN Number | UN1549 |
| Merck Index | 14,713 |
| Solubility Information | Insoluble in water. Soluble in concentrated acids. |
| Formula Weight | 339.68 |
| Percent Purity | 99.9% |
| Assay | (metals basis) |
| Chemical Name or Material | Antimony(III) sulfide |
Safety and Handling
| Hazard Category | H302-H312-H315-H319-H335-H500 |
| Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S. |
| EINECSNumber | 215-713-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only