missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) oxide, Puratronic™, 99.999% (metals basis)

CAS: 1309-64-4 | O3Sb2 | 291.52 g/mol

Supplier:  Thermo Scientific Chemicals 01064114

 View more versions of this product

Catalog No. AA1064114


Only null left
Explore available promotions

Description

Description

Antimony(III) oxide finds use as flame retardant wherein it synergistically acts with halogenated materials. It is employed in opacifying glasses, ceramics and enamels. In the preparation of optical and ruby glass, antimony trioxide serves as a bubble remover. Further, it is a useful catalyst in the production of polyethylene terephthalate (PET plastic) and the vulcanization of rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1550°C (sublimation)
656°C
1309-64-4
25 g
O3Sb2
Very slightly soluble in water; soluble in potassium hydroxide,hydrochloricacid and acetic acid; insoluble in organic solvents.
[O--].[O--].[O--].[Sb+3].[Sb+3]
291.52
Odorless
Antimony(III) oxide
White
Powder
99.99% (Metals basis)
MFCD00011214
GHPGOEFPKIHBNM-UHFFFAOYSA-N
diantimony(3+) trioxidandiide
291.5
5.67

Chemical Identifiers

1309-64-4
291.52
GHPGOEFPKIHBNM-UHFFFAOYSA-N
[O--].[O--].[O--].[Sb+3].[Sb+3]
O3Sb2
MFCD00011214
diantimony(3+) trioxidandiide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H351

EINECSNumber : 215-175-0

RTECSNumber : CC5650000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only