missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) Oxide, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. p-4939027
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1078360100 100 g Plastic bottle
M1078361000 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1078360100 Supplier Technidata Supplier No. 1.07836.0100
Only null left
For analysis

Chemical Identifiers

CAS 1309-64-4
Molecular Formula O3Sb2
Molecular Weight (g/mol) 291.52
MDL Number MFCD00011214
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
Synonym di-Antimony trioxide
IUPAC Name diantimony(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]

Specifications

Physical Form Solid
Quantity 100 g
Formula Weight 291.49
Odor Odorless
Packaging Plastic bottle
Appearance White solid
Vapor Pressure 13.3 hPa at 660°C
Chemical Name or Material Antimony(III) oxide
Hazard Category H351: Suspected of causing cancer.
EINECSNumber 215-175-0