missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) Oxide, Extra Pure, MilliporeSigma™

Catalog No. M1078352500
Change view
Click to view available options
Quantity:
2.5 kg
50 kg
Packaging:
Fibre carton
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1078352500 2.5 kg Plastic bottle
M1078359050 50 kg Fibre carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1078352500 Supplier Technidata Supplier No. 1.07835.2500

Chemical Identifiers

CAS 1309-64-4
Molecular Formula O3Sb2
Molecular Weight (g/mol) 291.52
MDL Number MFCD00011214
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
Synonym di-Antimony trioxide
IUPAC Name diantimony(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]

Specifications

Physical Form Solid
Quantity 2.5 kg
Formula Weight 291.49
Grade Extra Pure
Odor Odorless
Packaging Plastic bottle
Appearance White solid
Vapor Pressure 13.3 hPa at 660°C
Chemical Name or Material Antimony(III) oxide
Hazard Category H351: Suspected of causing cancer.