missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) oxide, 99%

Catalog No. p-7081233
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1112322 100 g
AAA1112336 500 g
AAA111230E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1112322 Supplier Thermo Scientific Chemicals Supplier No. A1112322
Only null left

CAS: 1309-64-4 | O3Sb2 | 291.52 g/mol

Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 1309-64-4
Molecular Formula O3Sb2
Molecular Weight (g/mol) 291.52
MDL Number MFCD00011214
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
IUPAC Name diantimony(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]

Specifications

Boiling Point 1550°C (sublimation)
Melting Point 656°C
Quantity 100 g
Merck Index 14,711
Solubility Information Insoluble in water.
Formula Weight 291.5
Percent Purity 99%
Odor Odorless
Density 5.67
Chemical Name or Material Antimony (III) oxide
Hazard Category H351
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 215-175-0
RTECSNumber CC5650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only