missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony(III) oxide, 99.6% min
Description
It finds applications in flame retardants, catalysts, batteries, and alloys. It acts as an opacifying agent in glasses, ceramics and enamels. It is used as a catalyst in the manufacture of PET plastic (polyethylene terephthalate), and the vulcanization of rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1550°C (sublimation) |
| Melting Point | 656°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 2 kg |
| Odor | Odorless |
| Appearance | White powder |
| Assay Percent Range | 99.6% min (Metal basis) |
| Density | 5.67 |
| Solubility Information | Poor solubility in water. Slowly dissolves in warm HCl or warm KOH solutions. |
| Show More |
Safety and Handling
| Hazard Category | H351 |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 215-175-0 |
| RTECSNumber | CC5650000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only