missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) oxide, 99.6% min

Catalog No. AA1157918
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1157918 50 g
AA1157936 500 g
AA11579A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. AA1157918 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' 01157918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1309-64-4 | O3Sb2 | 291.52 g/mol

It finds applications in flame retardants, catalysts, batteries, and alloys. It acts as an opacifying agent in glasses, ceramics and enamels. It is used as a catalyst in the manufacture of PET plastic (polyethylene terephthalate), and the vulcanization of rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1309-64-4
Molecular Formula O3Sb2
Molecular Weight (g/mol) 291.52
MDL Number MFCD00011214
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
Synonym Antimony trioxide
IUPAC Name diantimony(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]

Specifications

Boiling Point 1550°C (sublimation)
Melting Point 656°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.6% min (Metal basis)
Solubility Information Poor solubility in water. Slowly dissolves in warm HCl or warm KOH solutions.
Formula Weight 291.5
Odor Odorless
Appearance White powder
Density 5.67 g/cm3
Chemical Name or Material Antimony(III) oxide
Show More Show Less
missing translation for 'healthHazard1' H351
missing translation for 'healthHazard3' P201-P202-P281-P308+P313-P501c
EINECSNumber 215-175-0
RTECSNumber CC5650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only