Learn More
Antimony(III) ethoxide, 99.9% (metals basis)
CAS: 10433-06-4 | C6H15O3Sb | 256.943 g/mol
$154.82 - $1489.92
Chemical Identifiers
| CAS | 10433-06-4 |
|---|---|
| Molecular Formula | C6H15O3Sb |
| Molecular Weight (g/mol) | 256.943 |
| MDL Number | MFCD00015123 |
| InChI Key | JGOJQVLHSPGMOC-UHFFFAOYSA-N |
| Synonym | antimony triethoxide, triethoxyantimony, acmc-20alrx, sb oet 3, antimony iii ethoxide, antimony 3+ ; ethanolate, ethanol, antimony 3+ salt 9ci, antimony 3+ tris ethoxide, antimony 3+ ion tris ethoxide |
| PubChem CID | 66326 |
| IUPAC Name | antimony(3+);ethanolate |
| SMILES | CC[O-].CC[O-].CC[O-].[Sb+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4309706
|
Thermo Scientific Chemicals
04309706 |
5 g |
Each for $154.82
|
|
|||||
|
AA4309714
|
Thermo Scientific Chemicals
04309714 |
25 g |
Each for $568.42
|
|
|||||
|
AA4309722
|
Thermo Scientific Chemicals
04309722 |
100 g |
Each for $1,489.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10433-06-4 | |
| 256.943 | |
| JGOJQVLHSPGMOC-UHFFFAOYSA-N | |
| 66326 | |
| CC[O-].CC[O-].CC[O-].[Sb+3] |
| C6H15O3Sb | |
| MFCD00015123 | |
| antimony triethoxide, triethoxyantimony, acmc-20alrx, sb oet 3, antimony iii ethoxide, antimony 3+ ; ethanolate, ethanol, antimony 3+ salt 9ci, antimony 3+ tris ethoxide, antimony 3+ ion tris ethoxide | |
| antimony(3+);ethanolate |
Specifications
| 10433-06-4 | |
| 95°C (11 mmHg) | |
| C6H15O3Sb | |
| 5 g | |
| Moisture sensitive | |
| JGOJQVLHSPGMOC-UHFFFAOYSA-N | |
| antimony(3+);ethanolate | |
| 66326 | |
| 99.9% | |
| Antimony (III) ethoxide |
| 1.524 g/mL | |
| >110°C (230°F) | |
| MFCD00015123 | |
| UN3082 | |
| antimony triethoxide, triethoxyantimony, acmc-20alrx, sb oet 3, antimony iii ethoxide, antimony 3+ ; ethanolate, ethanol, antimony 3+ salt 9ci, antimony 3+ tris ethoxide, antimony 3+ ion tris ethoxide | |
| CC[O-].CC[O-].CC[O-].[Sb+3] | |
| 256.943 | |
| 256.9 | |
| Liquid | |
| (metals basis) |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 233-914-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only