missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) ethoxide, 99.9% (metals basis)

CAS: 10433-06-4 | C6H15O3Sb | 256.943 g/mol

Supplier:  Thermo Scientific Chemicals 04309714

 View more versions of this product

Catalog No. AA4309714

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Antimony (III) ethoxide
1.524 g/mL
>110°C (230°F)
C6H15O3Sb
25 g
Moisture sensitive
JGOJQVLHSPGMOC-UHFFFAOYSA-N
antimony(3+);ethanolate
66326
99.9%
10433-06-4
95°C (11 mmHg)
(metals basis)
MFCD00015123
UN3082
antimony triethoxide, triethoxyantimony, acmc-20alrx, sb oet 3, antimony iii ethoxide, antimony 3+ ; ethanolate, ethanol, antimony 3+ salt 9ci, antimony 3+ tris ethoxide, antimony 3+ ion tris ethoxide
CC[O-].CC[O-].CC[O-].[Sb+3]
256.943
256.9
Liquid

Chemical Identifiers

10433-06-4
256.943
JGOJQVLHSPGMOC-UHFFFAOYSA-N
66326
CC[O-].CC[O-].CC[O-].[Sb+3]
C6H15O3Sb
MFCD00015123
antimony triethoxide, triethoxyantimony, acmc-20alrx, sb oet 3, antimony iii ethoxide, antimony 3+ ; ethanolate, ethanol, antimony 3+ salt 9ci, antimony 3+ tris ethoxide, antimony 3+ ion tris ethoxide
antimony(3+);ethanolate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 233-914-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only